Learn More
HLA-DM Mouse anti-Human, PE, Clone: MAP.DM1, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 555983
Beschreibung
Reacts with human leukocyte antigen-DM (HLA-DM), a non-classical MHC class II molecule expressed in the cytoplasm of antigen presenting cells (APC). HLA-DM is composed of alpha and beta subunits which form a similar structure as the classical class II molecules. HLA-DM catalyzes the dissociation of CLIP from MHC class II-CLIP complexes in vitro and facilitates the binding of antigenic peptides. It also prevents self-antigens from becoming stably complexed with class II molecules and being presented to T cells.
Host Species: Mouse
Clone: MaP.DM1
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Spezifikation
| HLA-DM | |
| Monoclonal | |
| PE | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| MAP.DM1 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only.