Learn More
Invitrogen™ Human IgG Isotype Control
Human Polyclonal Isotype Control
Marke:  Invitrogen™ 12000C
Artikelnummer. 10537973
  • 240.00 € / 1 Milliliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.
Beschreibung
Beschreibung
Specificity was determined by ELISA using a panel of antibodies specific to human IgG, IgA, IgM, Kappa and Lambda. Concentration provided: 1 mg/mL. Antibody value assigned is based on the extinction coefficient of IgG: Optical Density at 280 nm of 1.4 equals 1 mg antibody.
Human IgG, also known as human immunoglobulin G, is an antibody that is produced by the immune system in response to antigens. It is the most abundant immunoglobulin in the blood and plays a key role in defense against pathogens. IgG works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG is also involved in other immune responses, such as antibody-dependent cellular cytotoxicity and neutralization of toxins. Human IgG can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Human IgG
Regelung
Unconjugated
Immunoglobulin G; ImmunoglobulinG
Gamma Immunoglobin
1 mL
Isotype Control (Primary)
-20°C
Liquid
Responsibly packaged
Control, Flow Cytometry
1 mg/mL
PBS with 0.1% sodium azide
Human
Protein G
RUO
Not Applicable
Isotype Control