Learn More
Invitrogen™ Donkey anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 488
Donkey Polyclonal Secondary Antibody
Marke:  Invitrogen™ SA510126
Artikelnummer. 13477277
  • 208.00 € / 500 Mikrogramm
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
This antibody is cross-adsorbed and exhibits minimum reactivity to bovine, chicken, goat, mouse, rabbit, rat and sheep.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Human IgG (H+L) Cross-Adsorbed, Human IgG (H+L)
Polyclonal
DyLight 488
Donkey
Antigen affinity chromatography
RUO
Human
Secondary Antibody
IgG
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4
Human IgG-heavy and light chain.
500 μg
Secondary
4°C
Liquid
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen