missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza B HA Monoclonal Antibody (B114), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA516290
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Influenza A and B are the two types of influenza viruses that cause epidemic human disease. Influenza B viruses currently circulating are divided into two antigenically and genetically distinct groups: the Victoria and Yamagata lineages. Influenza virus B is a genus in the virus family Orthomyxoviridae. A limited host range means that Influenza virus B pandemics are rare. The nucleoprotein protects the negative strand viral RNA from nucleases by encapsidating it. Encapsidated genomic RNA is termed the ribonucleoprotein (RNP) and serves as template for transcription and replication.
Spezifikation
| Influenza B HA | |
| Monoclonal | |
| 6.41 mg/mL | |
| PBS with 15mM sodium azide; pH 7.5 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2b |
| Immunocytochemistry | |
| B114 | |
| Unconjugated | |
| flu hemagglutinin; Influenza B; influenza b hemagglutinin; Influenza HA; influenza hemagglutinin | |
| B/Lee/40 and B/Singapore/-222/79 virus | |
| 2 mg | |
| Primary | |
| 4° C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur