missing translation for 'onlineSavingsMsg'
Learn More

IQGAP1 Mouse, Unlabeled, Clone: 24, BD

Artikelnummer. p-3856104
Change view
Click to view available options
Menge:
150 μg
50 μg
Unit Size:
1 Set
Product Code. Menge unitSize
15885539 50 μg 1 Set
15895539 150 μg 1 Set
2 options
This item is not returnable. View return policy

Product Code. 15885539

Brand: BD Biosciences 610611

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GTPases (Ras, Rho, cdc42Hs, and rac) modulate various cellular functions such as cytoskeletal architecture, growth, motility, and gene expression. The activity of the GTP-binding proteins is regulated by factors that accelerate GTP-binding (GAPs) and proteins that enhance the rate of GTP hydrolysis (GTPases). IQGAP1 is Ras-GAP related protein that contains a WW domain that binds poly-proline regions, a calponin-homology domain (CHD), an IQ domain of unconventional myosins, and a C-terminal GAP-related domain (GRD). IQGAP1 co-immunoprecipitates with cdc42 and calmodulin, and colocalizes with cdc42Hs at the leading edges of the plasma membrane and at the cell-cell junctions. IQGAP1 may interact with both cadherins and catenins leading to dissociation of these proteins from sites of cell adhesion. Thus, IQGAP1 may be a Rac1 and Cdc42 effector that negatively regulates cadherin-mediated cell junctions.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IQGAP1
Anwendungen Western Blot
Klassifikation Monoclonal
Klon 24
Konzentration 250μg/mL
Konjugat Unconjugated
Zusammensetzung Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Wirtsspezies Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Reinigungsverfahren Affinity Purified
Menge 50 μg
Regulatorischer Status RUO
Forschungsgebiet Cell Biology
Primär oder sekundär Primary
Zielspezies Canine, Human, Mouse, Rat
Inhalt und Lagerung Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.