missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Laminin beta-3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA521514
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Recommended positive controls: A431, NIH-3T3, mouse bone marrow. Predicted reactivity: Mouse (81%), Rat (80%), Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The product encoded by this gene is a laminin that belongs to a family of basement membrane proteins. This protein is a beta subunit laminin, which together with an alpha and a gamma subunit, forms laminin-5. Mutations in this gene cause epidermolysis bullosa junctional Herlitz type, and generalized atrophic benign epidermolysis bullosa, diseases that are characterized by blistering of the skin. Multiple alternatively spliced transcript variants that encode the same protein have been found for this gene.
Spezifikation
| Laminin beta-3 | |
| Polyclonal | |
| Unconjugated | |
| LAMB3 | |
| AI1A; BM600-125KDA; Epiligrin subunit bata; kalinin B1 chain; Kalinin subunit beta; kalinin-140kDa; LAM5; Lamb3; Laminin B1k chain; laminin beta 3; laminin chain; laminin S B3 chain; laminin subunit beta 3; laminin subunit beta-3; laminin, beta 3; laminin, beta 3 (nicein (125kD), kalinin (140kD), BM600 (125kD)); laminin-5 subunit beta; LAMNB1; Nicein subunit beta; nicein, 125kD; nicein, 125kDa; nicein-125k; nicein-125kDa; RP1-272L16.3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16780, 3914 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.46 mg/mL | |
| PBS with 20% glycerol, 1% BSA and 0.025% ProClin 300; pH 7 | |
| Q13751, Q61087 | |
| LAMB3 | |
| Recombinant protein encompassing a sequence within the center region of human Laminin beta 3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur