missing translation for 'onlineSavingsMsg'
Learn More

LT-α Mouse anti-Human, Unlabeled, Clone: 359-238-8, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 551222

Product Code. 15816578

  • ONLINE EXCLUSIVE PRICE
    435.00 €
    434.42 € / 1 Stück
    Sparen 0.58 €
    0.13% Rabatt

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

Description

Description

The 359-238-8 antibody reacts with human lymphotoxin-α (LT-α, formerly known as TNF-β). The immunogen used to generate the 359-238-8 hybridoma was recombinant human LT-α. This is a neutralizing antibody.

Host Species: Mouse
Clone: 359-238-8
Isotype: IgG1
Species Reactivity: Human
Immunogen: Recombinant human LT-α

ELISA

TRUSTED_SUSTAINABILITY
Specifications

Specifications

LT-α
Monoclonal
1.0mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Recombinant human LT-α
1 mg
Immunology
Human
IgG1
ELISA
359-238-8
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.