Learn More
IgG (H+L) F(ab')2-Goat anti-Mouse, NANOGOLD, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
Marke: Invitrogen N24915
Beschreibung
The NANOGOLD Fab' fragment of goat anti-mouse IgG is covalently conjugated with uniform, 1.4 nm-diameter gold clusters. When used with our LI Silver Enhancement Kit (L-24919), the NANOGOLD conjugates can be used as a probe in immunoblotting, light microscopy or electron microscopy.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation
| Mouse IgG (H+L) | |
| Polyclonal | |
| Nanogold | |
| Gamma Immunoglobin | |
| 1 mL | |
| Secondary | |
| 4° C | |
| IgG |
| Immunomicroscopy | |
| 0.08 mg/mL | |
| Goat | |
| Purified | |
| RUO | |
| Mouse | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.