missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Mouse IgG1 Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 594

Artikelnummer. 10357212
Change view
Click to view available options
Menge:
500 μg
Packungsgröße:
250 Mikrogramm
Artikelnummer. Menge unitSize
10357212 500 μg 250 Mikrogramm
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Artikelnummer. 10357212

Marke: Invitrogen™ A21125

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Goat Polyclonal Secondary Antibody

To minimize cross-reactivity, these goat anti-mouse IgG1 whole secondary antibodies have been affinity purified and cross-adsorbed against mouse IgM, mouse IgA, pooled human sera, purified human paraproteins, and mouse isotypes IgG2a, IgG2b, and IgG3 prior to conjugation. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in higher sensitivity and less background staining. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondaries flow through. The benefits of this extra step are apparent in multiplexing/multicolor-staining experiments (e.g., flow cytometry) where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there are may be the presence of endogenous immunoglobulins. Alexa Fluor dyes are among the most trusted fluorescent dyes available today. Invitrogen™ Alexa Fluor 594 dye is a bright, red-fluorescent dye with excitation ideally suited to the 594 nm laser line. For stable signal generation in imaging and flow cytometry, Alexa Fluor 594 dye is pH-insensitive over a wide molar range. Probes with high fluorescence quantum yield and high photostability allow detection of low-abundance biological structures with great sensitivity. Alexa Flu...

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Mouse IgG1 Cross-Adsorbed
Anwendungen Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Konzentration 2 mg/mL
Konjugat Alexa Fluor 594
Zusammensetzung PBS with 5mM sodium azide; pH 7.5
Wirtsspezies Goat
Immunogen IgG gamma 1.
Reinigungsverfahren Purified
Menge 500 μg
Regulatorischer Status RUO
Primär oder sekundär Secondary
Zielspezies Mouse
Inhalt und Lagerung 4°C, store in dark
Produkttyp Secondary Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.