Learn More
Nestin Mouse, Unlabeled, Clone: 25/NESTIN, BD
Artikelnummer. p-3850342
Change view
Click to view available options
Menge:
150 μg
50 μg
Packungsgröße:
1 Set
1 Stück
Artikelnummer. Menge unitSize
15895999 50 μg 1 Set
10308469 150 μg 1 Stück
2 options
Artikelnummer. 15895999
Marke: BD Biosciences 611658
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody
The cytoskeleton consists primarily of core structural proteins that include microfilaments, microtubules, and intermediate filaments (IFs). IFs contain more than 50 distinct proteins that are organized into six different subtypes: Type I/II keratin proteins expressed in epithelia cells, type III vimentin/desmin, type IV neurofilament proteins, type V nuclear lamins, and type VI nestin proteins expressed primarily in embryonic cells. Nestin has a conserved core region (amino acids 7 to 314), which contains an α helical domain that is involved in coiled-coil assembly of IFs. The C-terminal region of nestin is similar to type IV IFs, since it contains highly charged amino acids, many glutamate residues, and an 11 amino acid repeat motif. Nestin is expressed in the cerebrum during embryonic development, in the cerebellum during early postnatal development, and in dermatomal cells and myoblasts during myogenesis. In vitro, nestin forms homodimers and homotetramers, but not IFs, and can co-assemble with type III vimentin and type IV internexin proteins. Thus, nestin is a core IF protein that is essential for proper cytoskeletal formation during neurogenesis and myogenesis. Nestin molecular weight can vary anywhere from 200 to 260kDa depending on where it is being expressed.
Host Species: Mouse
Clone: 25
Isotype: IgG1, κ
Species Reactivity [for Features Main]: Rat
Immunogen: Rat Nestin aa. 402-604
Immunofluorescence, Western Blotting
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Nestin
Anwendungen Western Blot
Klassifikation Monoclonal
Klon 25/NESTIN
Konzentration 250μg/mL
Konjugat Unconjugated
Zusammensetzung Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Wirtsspezies Mouse
Immunogen Rat Nestin aa. 402-604 Recombinant Protein
Reinigungsverfahren Affinity Purified
Menge 50 μg
Regulatorischer Status RUO
Forschungsgebiet Cell Biology
Primär oder sekundär Primary
Zielspezies Human, Rat
Inhalt und Lagerung Store undiluted at -20°C.
Isotype IgG1 κ