Learn More
Oct3/4 Mouse, Unlabeled, Clone: 40/OCT-3, BD
Mouse Monoclonal Antibody
178.69 € - 339.19 €
Spezifikation
| Antigen | OCT3/4 |
|---|---|
| Klon | 40/OCT-3 |
| Konzentration | 250μg/mL |
| Anwendungen | In vitro Assay, Western Blot |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
15845839
|
BD Biosciences
611202 |
50 μg |
349.00 € 178.69 € / 1 Set Sparen 170.31 € 48% Rabatt |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
|
15855839
|
BD Biosciences
611203 |
150 μg |
340.00 € 339.19 € / 1 Set Sparen 0.81 € 0.24% Rabatt |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
Beschreibung
Development of a multicellular organism from a single fertilized cell is regulated by the coordinated activity of DNA transcription factors. Oct3/4, a member of the POU family of transcription factors, functions in pluripotent cells of early embryonic stem cell (ES) lines and embryonal carcinomas (EC). Other members of the POU family include Oct1, Oct2, Pit-1, and unc-86. The POU domain, a 150-amino acid region that determines binding specificity, is conserved among these proteins and consists of 3 subdomains: POU-specific A and B
subdomains and a homeobox-like subdomain. Oct3/4 is expressed in undifferentiated cells, but is lost as cells are induced to differentiate. Oct3/4 is not expressed in adult tissues. The interaction of Oct3/4 with SOX2, another embryonic transcription factor, produces an active complex that regulates expression of genes such as Nanog, UTF1, and FGF4. Although Oct3/4 is specifically phosphorylated on serine residues, this modification is not required for DNA binding, but may affect its transactivation potential. Thus, Oct3/4 is a transcription factor that plays an important role in determining early steps of embryogenesis and differentiation.
Host Species: Mouse
Clone: 40
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Mouse Oct3 aa. 252-372 Recombinant Protein
Formula Weight [Chemical]: 46kDa
Western Blotting
Spezifikation
| OCT3/4 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| Oct3, OTF3, Oct4, OTF4, POU5F1 |
| 40/OCT-3 | |
| In vitro Assay, Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Mouse Oct3 aa. 252-372 Recombinant Protein | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.