Learn More
p21 protein (Cdc42/Rac)-activated kinase 1, Mouse, Clone: 1E11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PAK1.
Marke: Abnova H00005058-M01.100ug
Beschreibung
PAK proteins are critical effectors that link RhoGTPases to cytoskeleton reorganization and nuclear signaling. PAK proteins, a family of serine/threonine p21-activating kinases, include PAK1, PAK2, PAK3 and PAK4. These proteins serve as targets for the small GTP binding proteins Cdc42 and Rac and have been implicated in a wide range of biological activities. PAK1 regulates cell motility and morphology. Alternativelt spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: APRPEHTKSVYTRSVIEPLPVTPTRDVATSPISPTENNTTPPDALTRNTEKQKKKPKMSDEEILEKLRSIVSVGDPKKKYTRFEKIGQGASpezifikation
| p21 protein (Cdc42/Rac)-activated kinase 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_002576 | |
| PAK1 | |
| PAK1 (NP_002567, 191 a.a. ∼ 280 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Apoptosis | |
| 5058 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 1E11 | |
| Mouse monoclonal antibody raised against a partial recombinant PAK1. | |
| PAK1 | |
| MGC130000/MGC130001/PAKalpha | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.