Learn More
twinfilin, actin-binding protein, homolog 1 (Drosophila), Mouse, Clone: 1E2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PTK9.
Marke: Abnova H00005756-M02.100ug
Beschreibung
This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1. [provided by RefSeq
Sequence: VAFPISREAFQALEKLNNRQLNYVQLEIDIKNEIIILANTTNTELKDLPKRIPKDSARYHFFLYKHSHEGDYLESIVFIYSMPGYTCSIRERMLYSSCKSRLLEIVERQLQMDVIRKIEIDNGDELTADFLYEEVHPKQHAHKQSSAKPKGPAGKRGIRRLIRGPAETEATTDSpezifikation
| twinfilin, actin-binding protein, homolog 1 (Drosophila) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC022344 | |
| TWF1 | |
| PTK9 (AAH22344, 80 a.a. ∼ 252 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Kinases | |
| Primary | |
| Human, Rat | |
| Antibody |
| ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| 1E2 | |
| Mouse monoclonal antibody raised against a partial recombinant PTK9. | |
| TWF1 | |
| A6/MGC23788/MGC41876/PTK9 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 5756 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.