missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Salmonella typhimurium LPS Monoclonal Antibody (1E6)

Beschreibung
MA1-22693 detects Salmonella typhimurium 0 - 4 in bacteria samples.
The genus Salmonella is a member of the family Enterobacteriaceae. The genus is composed of Gram negative bacilli that are facultative and flagellated (motile). Salmonellae possess 3 major surface antigens; the H or flagellar antigen (phase 1 and 2), the O or somatic antigen (part of the LPS moiety) and the Vi or capsular antigen (referred to as K in other Enterobacteriaceae). Salmonellae also possess the LPS endotoxin characteristic of Gram negative bacteria. This LPS is composed of an O polysaccharide (O antigen) an R core and the endotoxic inner Lipid A. Endotoxins evoke fever and can activate complement, kinin and clotting factors.
Specifications
Specifications
| Antigen | Salmonella typhimurium LPS |
| Anwendungen | Dot Blot, ELISA, Flow Cytometry, Immunocytochemistry |
| Klassifikation | Monoclonal |
| Klon | 1E6 |
| Konzentration | 4.8 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 0.09% sodium azide; pH 7.4 |
| Gen-Alias | S. typhimurium; Salmonella LPS; Salmonellae |
| Wirtsspezies | Mouse |
| Immunogen | Lipopolysaccharides (LPS) of S. typhimurium. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?