Learn More
TRAF3 interacting protein 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Beschreibung
This gene encodes a protein involved in regulating responses to cytokines by members of the Rel/NF-kappaB transcription factor family. These factors play a central role in innate immunity in response to pathogens, inflammatory signals and stress. This gene product interacts with TRAF proteins (tumor necrosis factor receptor-associated factors) and either I-kappaB kinase or MAP kinase to activate either NF-kappaB or Jun kinase. Several alternative transcripts encoding different isoforms have been identified. Another transcript, which does not encode a protein and is transcribed in the opposite orientation, has been identified. Overexpression of this transcript has been shown to reduce expression of at least one of the protein encoding transcripts, suggesting it has a regulatory role in the expression of this gene. [provided by RefSeq
Spezifikation
Spezifikation
| Antigen | TRAF3 interacting protein 2 |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Beschreibung | Rabbit polyclonal antibody raised against a full-length human TRAF3IP2 protein. |
| Zusammensetzung | PBS with no preservative; pH 7.4 |
| Gen | TRAF3IP2 |
| Gen-Zugriffsnummer | NM_147686 |
| Gen-Alias | ACT1/C6orf2/C6orf4/C6orf5/C6orf6/CIKS/DKFZp586G0522/MGC3581 |
| Gensymbole | TRAF3IP2 |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.