missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Antibody Labeling Kits for 100 μg

Artikelnummer. 30282834
Ansicht ändern
Click to view available options
Marker oder Farbstoff:
Alexa Fluor 594
Alexa Fluor 350
Alexa Fluor 488
Alexa Fluor 532
Alexa Fluor 546
Alexa Fluor 555
Alexa Fluor 568
Alexa Fluor 647
Alexa Fluor 680
Alexa Fluor 790
Biotin
Menge:
3 x 100 μg
Packungsgröße:
1 Stück
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Marker oder Farbstoff Menge Packungsgröße
30282834 Biotin 3 x 100 μg 1 Stück
30283372 Alexa Fluor 350 3 x 100 μg 1 Stück
30283944 Alexa Fluor 488 3 x 100 μg 1 Stück
30283961 Alexa Fluor 532 3 x 100 μg 1 Stück
30282686 Alexa Fluor 546 3 x 100 μg 1 Stück
30283622 Alexa Fluor 555 3 x 100 μg 1 Stück
30283001 Alexa Fluor 568 3 x 100 μg 1 Stück
30283956 Alexa Fluor 594 3 x 100 μg 1 Stück
30283613 Alexa Fluor 647 3 x 100 μg 1 Stück
30284082 Alexa Fluor 680 3 x 100 μg 1 Stück
30283741 Alexa Fluor 790 3 x 100 μg 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30282834 Lieferant Invitrogen™ Lieferanten-Nr. B88060
 Eingeschränkter Artikel
Lager vorrätig
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Alexa Fluor™ Antibody Labeling Kits are designed for efficiency and reliability, delivering ready-to-use labeled proteins in just 30 minutes, with minimal hands-on time. The kits have been redeveloped with user-friendly, advanced purification columns that ensure rapid dye removal and high yield of pure conjugated antibody.

Alexa Fluor antibody labeling kits are designed for efficiency and reliability, delivering ready-to-use labeled proteins in just 30 minutes, with minimal hands-on time. The kits have been redeveloped with user-friendly, advanced purification columns that ensure rapid dye removal and high yield of pure conjugated antibody.

Alexa Fluor antibody labeling kit features include:

  • Fast and convenient—labeled proteins ready for use in 30 minutes, with less than 15 minutes of hands-on time
  • High efficiency—label 20 to 100 μg of protein with bright, photostable Alexa Fluor dyes, with three reactions per kit
  • Exceptional recovery—pre-packed spin columns for rapid removal of unbound dye and high conjugate yield
  • Comprehensive guidance—includes detailed instructions and a convenient calculator for determining the degree of labeling (DOL)
  • Versatile applications—suitable for labeling a variety of purified proteins, including antibodies, growth factors, cytokines, nanobodies, and enzymes

Superior performance and a faster workflow

Alexa Fluor antibody labeling kits deliver superior results with lower background fluorescence, reduced nonspecific binding, and streamlined workflows. Each kit is equipped with materials for three antibody conjugation reactions with one of ten different fluorophores or biotin. Bright, photostable Alexa Fluor dyes react efficiently with primary amines of proteins to form stable dye–protein conjugates. Removal of free dye with the included spin columns enables accurate determination of dye‐to‐protein ratios.

The Alexa Fluor antibody labeling protocol is simple and rapid, ensuring minimal hands-on time and reliable results. The kits include all necessary reagents for antibody labeling, including amine-reactive dye, buffer, and purification columns. The reactive dyes contained in these kits readily react with lysine residues in antibodies or proteins to yield a covalently attached fluorescence-based sensor. Spin columns included in the kits are used for purifying the labeled antibody from excess dye with yields of 70–95%. The kits contain sufficient reagents for three labeling reactions of 20 to 100 μg of antibody. The wide variety of colors are compatible with various detection systems, including fluorescence microscopy, flow cytometry, and high content imaging platforms.

TRUSTED_SUSTAINABILITY

Spezifikation

Zur Verwendung mit (Anwendung) Antibody and Protein Labeling
Produkttyp Labeling Kit
Menge 3 x 100 μg
Nachweisverfahren Fluorescence
Produktlinie Alexa Fluor
Chemische Reaktivität Amine
Löslichkeit Aqueous Buffer, DMSO (Dimethylsulfoxide)
Marker oder Farbstoff Biotin
Labeling Target Antibodies, Proteins
Markertyp Biotin

For Research Use Only.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.