missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
AP2 alpha Monoclonal specifically detects AP2 alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Radioimmunoassay, CyTOF-ready.
Spezifikation
Specifications
| Antigen | AP2 alpha |
| Anwendungen | Western Blot |
| Klassifikation | Monoclonal |
| Klon | A6/2/2 |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000, CyTOF-ready |
| Gen-Alias | Activating enhancer-binding protein 2-alpha, Activator protein 2, AP-2 transcription factor, AP2-alpha, AP2TFAP-2alpha, AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha), BOFS, FLJ51761, TFAP2, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2-alpha |
| Gensymbole | TFAP2A |
| Wirtsspezies | Mouse |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?