missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aquaporin 0 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5114870
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Antibody detects endogenous levels of total AQP0.
AQP0 is the most abundant endogenous protein in the plasma membrane of lens fiber cells where it functions not only as a water pore, but it is also involved in fiber-fiber adhesion and is crucial for fiber cell structure and organization. AQP0 contains an additional pore constriction, not seen in any other aquaporin structures, which may be responsible for pore gating. The closed AQP0 pore holds just three water molecules, which are spaced too far apart to form hydrogen bonds with each other. The C-terminal domain of AQP0 undergoes extensive post-translational modification, including many truncations, during lens aging due to the actions of m-calpain, proteases or non-enzymatic mechanisms. These truncation sites may be involved in the development of cataracts.
Spezifikation
| Aquaporin 0 | |
| Polyclonal | |
| Unconjugated | |
| MIP | |
| AQP0; aquaporin 0; Aquaporin0; aquaporin-0; Cat; CTRCT15; dominant cataract; Hfi; hydropic fibers; lens fiber major intrinsic protein; lens opacity; LIM1; Lop; major intrinsic protein of eye lens fiber; major intrinsic protein of lens fiber; MIP; MIP26; MP26; Palm; shrivelled; Svl | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17339, 25480, 4284 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P09011, P30301, P51180 | |
| MIP | |
| A synthesized peptide derived from human MIP(Accession P30301), corresponding to amino acid residues V125-G175. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur