missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Aquaporin-4 Monoclonal antibody specifically detects Aquaporin-4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Aquaporin-4 |
| Anwendungen | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | CL0178 |
| Konjugat | DyLight 594 |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | aquaporin 4, MGC22454 |
| Wirtsspezies | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from P55087, with the exact immunogen sequence remaining proprietary. |
| Reinigungsverfahren | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?