Learn More
Invitrogen™ ARFGEF2 Recombinant Rabbit Monoclonal Antibody (24GB6840)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA556970
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
ARFGEF2 Recombinant Monoclonal Antibody for Western Blot
ADP-ribosylation factors (ARFs) play an important role in intracellular vesicular trafficking. The protein encoded by this gene is involved in the activation of ARFs by accelerating replacement of bound GDP with GTP and is involved in Golgi transport. It contains a Sec7 domain, which may be responsible for its guanine-nucleotide exchange activity and also brefeldin A inhibition. [provided by RefSeq, Jul 2008].
Spezifikation
| ARFGEF2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ARFGEF2 | |
| ADP ribosylation factor guanine nucleotide exchange factor 2; ADP-ribosylation factor guanine nucleotide-exchange factor 2; ADP-ribosylation factor guanine nucleotide-exchange factor 2 (brefeldin A-inhibited); AI463430; Arfgef2; ARFGEP2; AW493672; Big2; Brefeldin A-inhibited GEP 2; Brefeldin A-inhibited guanine nucleotide-exchange factor 2; brefeldin A-inhibited guanine nucleotide-exchange protein 2; dJ1164I10.1; E230011G24Rik; PVNH2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10564, 296380, 99371 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB6840 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A2A5R2, Q7TSU1, Q9Y6D5 | |
| ARFGEF2 | |
| A synthesized peptide derived from human ARFGEF2 (aa 1501-1514). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |