Learn More
Arginase 1/ARG1/liver Arginase Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 9 publications
250.00 €
Spezifikation
| Antigen | Arginase 1/ARG1/liver Arginase |
|---|---|
| Konzentration | 1 mg/mL |
| Verdünnung | Western Blot 1:1000-1:50000, Simple Western, Flow Cytometry 5 - 10 μg/mL, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:10 - 1:500, Knockdown Validated |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18515020
|
Novus Biologicals
NBP1-32731SS |
0.025 mg |
250.00 €
25 Mikrogramm |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
Beschreibung
Arginase 1/ARG1/liver Arginase Polyclonal specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.Spezifikation
| Arginase 1/ARG1/liver Arginase | |
| Western Blot 1:1000-1:50000, Simple Western, Flow Cytometry 5 - 10 μg/mL, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:10 - 1:500, Knockdown Validated | |
| Polyclonal | |
| Rabbit | |
| Cellular Markers, Chromatin Research, Lipid and Metabolism | |
| PBS with 0.02% Sodium Azide | |
| arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, Liver-type arginase, Type I arginase | |
| ARG1 | |
| IgG | |
| Affinity Purified | |
| 35 kDa |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| P05089 | |
| 383 | |
| Arginase 1/ARG1/liver Arginase Antibody made from full length human Arginase 1 Recombinant protein. | |
| Primary | |
| Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles. |