missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 13822-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody recognizes endogenous ARP3 and may cross-react with ATCR3B.
The specific function of this gene has not yet been determined; however, the protein it encodes is known to be a major constituent of the ARP2/3 complex. This complex is located at the cell surface and is essential to cell shape and motility through lamellipodial actin assembly and protrusion.Spezifikation
| ARP3 | |
| Polyclonal | |
| Unconjugated | |
| ACTR3 | |
| Actin like protein 3, Actin related protein 3, ACTR3, ACTR3B, Arp11, ARP3, Arp4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10096, 74117, 81732 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P61158, Q4V7C7, Q99JY9 | |
| Actr3 | |
| ARP3/ARP3B Fusion Protein Ag4922 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur