missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATF3 Recombinant Rabbit Monoclonal Antibody (24GB6925)

Rabbit Recombinant Monoclonal Antibody
146.00 € - 500.00 €
Spezifikation
| Antigen | ATF3 |
|---|---|
| Klon | 24GB6925 |
| Inhalt und Lagerung | -20°C |
| Anwendungen | Western Blot |
| Klassifikation | Recombinant Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
30275102
![]() |
Invitrogen™
MA556973 |
100 μL |
500.00 €
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30276552
|
Invitrogen™
MA556974 |
20 μL |
146.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
ATF3 Recombinant Monoclonal Antibody for Western Blot
ATF3 (Activating transcription factor 3) is a member of the mammalian activation transcription factor/cAMP responsive element-binding (CREB) protein family of transcription factors. It encodes a protein with a calculated molecular mass of 22 kD. ATF3 represses rather than activates transcription from promoters with ATF binding elements. An alternatively spliced form of ATF3 (ATF3 delta Zip) encodes a truncated form ATF3 protein lacking the leucine zipper protein-dimerization motif and does not bind to DNA. In contrast to ATF3, ATF3 delta Zip stimulates transcription presumably by sequestering inhibitory co-factors away from the promoter. It is possible that alternative splicing of the ATF3 gene may be physiologically important in the regulation of target genes. Three transcript variants encoding two different isoforms have been found for this gene.Spezifikation
| ATF3 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| ATF3 | |
| activating transcription factor 3; ATF 3; ATF3; cAMP-dependent transcription factor ATF-3; cyclic AMP-dependent transcription factor ATF-3; Liver regeneration factor 1; Lrf1; LRF-1; LRFI; LRG-21; MGC137608 protein; transcription factor LRG-21; zgc:55526 | |
| ATF3 | |
| Primary | |
| Affinity chromatography |
| 24GB6925 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P18847, Q60765 | |
| 11910, 467 | |
| Recombinant protein of human ATF3 (aa 1-181). | |
| Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts