Learn More
Invitrogen™ ATP6V0D1 Recombinant Rabbit Monoclonal Antibody (24GB6855)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA556985
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
ATP6V0D1 Recombinant Monoclonal Antibody for Western Blot
This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c'', and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is known as the D subunit and is found ubiquitously.
Spezifikation
| ATP6V0D1 | |
| Recombinant Monoclonal | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P51863, P61421 | |
| ATP6V0D1 | |
| Recombinant protein of human ATP6V0D1 (aa 1-351). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB6855 | |
| Unconjugated | |
| ATP6V0D1 | |
| 32 kd accessory protein; 32 kDa accessory protein; Ac39; AI267038; Atp6d; ATP6DV; Atp6v0d1; ATPase H+ transporting V0 subunit d1; ATPase, H+ transporting, lysosomal (vacuolar proton pump), 42 kDa; ATPase, H+ transporting, lysosomal (vacuolar proton pump), member D; ATPase, H+ transporting, lysosomal 38kDa, V0 subunit D; ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d1; ATPase, H+ transporting, lysosomal V0 subunit D1; ATPase, H+ transporting, lysosomal, V0 subunit d1; ATPase, H+ transporting, V0 subunit D; cto; fb73h07; FLJ43534; H(+)-transporting two-sector ATPase, subunit D; lysosomal 38kDa; P39; Physophilin; Unknown (protein for MGC:63769); Vacuolar proton pump subunit d 1; vacuolar proton-ATPase, subunit D; V-ATPase, subunit D; V-ATPase 40 KDa accessory protein; V-ATPase AC39 subunit; V-ATPase subunit d 1; V-ATPase, subunit D; VATX; Vma6; VPATPD; V-type proton ATPa; V-type proton ATPase subunit d 1; wu:fb73h07; zgc:63769 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11972, 291969, 9114 | |
| -20°C | |
| Liquid |