Learn More
Invitrogen™ AVIL Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA590703
Beschreibung
Immunogen sequence: AAVQVRVRMG TEPRHFMAIF KGKLVIFEGG TSRKGNAEPD PPVRLFQIHG NDKSNTKAVE VPAFASSLNS NDVFLLRTQA EHYLWYGKGS SGDERAMAKE LASLLCDGSE NTVAEGQEPA EFWDLLGGKT PYANDKRLQQ EILDVQSRLF ECSNKTGQFV VTEITDFTQD DLNPTDVMLL DTWDQVFLWI GAEANATEKE SALATAQQYL HTHPSGRDPD TPILIIKQGF EPPIFTGWFL AWDPNIWSAG KTYEQLKEEL GDAAAIMRIT ADMKNATLSL NSNDSEPKYY PIAVLLKNQN QELPEDVNPA KKENYLSEQD FVSVFGITRG QFAALPGWKQ LQMKKEKGLF; Positive Samples: HT-29, Mouse uterus, Mouse brain, Rat uterus, Rat testis, Rat brain; Cellular Location: Cell projection, Cytoplasm, axon, cytoskeleton.
AVIL is a member of the gelsolin/villin family of actin regulatory proteins. This protein has structural similarity to villin. It binds actin and may play a role in the development of neuronal cells that form ganglia.
Spezifikation
| AVIL | |
| Polyclonal | |
| Unconjugated | |
| AVIL | |
| actin-binding protein DOC6; Advil; Advillin; Advillin-like protein; Avil; DOC6; p92; peripheral nervous system villin-like protein; pervin; Unknown (protein for MGC:136857); zgc:136857 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10677, 11567, 79253 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.45 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O75366, O88398, Q9WU06 | |
| AVIL | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.