missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZD7687, MedChemExpress
Beschreibung
AZD7687 is a potent, selective, reversible and orally active diacylglycerol acyltransferase 1 (DGAT1) inhibitor with an IC50 of 80 nM for human DGAT1. AZD7687 can be used for type 2 diabetes mellitus and obesity research.
Spezifikation
Spezifikation
| Chemischer Name oder Material | AZD7687 |
| CAS | 1166827-44-6 |
| Farbe | Pink |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C21H25N3O3 |
| Menge | 10 mg |
| Löslichkeitsinformationen | DMSO : ≥ 50 mg/mL (136.08 mM) |
| SMILES | CC1=C(C(N)=O)N=C(C2=CC=C([C@H]3CC[C@H](CC(O)=O)CC3)C=C2)C(C)=N1 |
| Molekulargewicht (g/mol) | 367.44 |
| Formelmasse | 367.44 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?