Learn More
b-Methylamino-L-alaninhydrochlorid, TRC
Specifications
Specifications
| Chemischer Name oder Material | beta-Methylamino-L-alanine Hydrochloride |
| CAS | 16012-55-8 |
| Summenformel | C4H10N2O2 • (HCl) |
| Menge | 50 mg |
| Synonym | 2-Amino-3-(methylamino)propanoic Acid Hydrochloride, S(+)-β-Methyl-α,β-diaminopropionic Acid Hydrochloride, BMAA Hydrochloride |
| InChI-Formel | InChI=1S/C4H10N2O2.ClH/c1-6-2-3(5)4(7)8;/h3,6H,2,5H2,1H3,(H,7,8);1H/t3-;/m0./s1 |
| SMILES | CNC[C@H](N)C(O)=O.Cl |
| IUPAC-Name | (2S)-2-Amino-3-(methylamino)propansäure;hydrochlorid |
| Molekulargewicht (g/mol) | 118.133646 |
| Formelmasse | 154.0509 |