missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
B7-H7/HHLA2 Monoclonal antibody specifically detects B7-H7/HHLA2 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
Specifications
Specifications
| Antigen | B7-H7/HHLA2 |
| Anwendungen | Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF |
| Klassifikation | Monoclonal |
| Klon | 907812R |
| Konjugat | DyLight 488 |
| Verdünnung | Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | HERV-H LTR-associating 2, HERV-H LTR-associating protein 2, Human endogenous retrovirus-H long terminal repeat-associating protein 2 |
| Wirtsspezies | Mouse |
| Immunogen | Human embryonic kidney cell line HEK293-derived human B7-H7/HHLA2, Met1-Asn344, Accession # Q9UM44 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?