Learn More
B7-H7 Monoclonal Antibody (MA57YW), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen 17-6537-41
Beschreibung
Description: This MA57YW monoclonal antibody recognizes human B7-H7 on live and paraformaldehyde-fixed cells. Applications Reported: This MA57YW antibody has been reported for use in flow cytometric analysis. Applications Tested: This MA57YW antibody has been pre-diluted and tested by flow cytometric analysis of the HDLM2 cell line. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser
B7-H7 Monoclonal antibody specifically detects B7-H7 in Human samples. It is validated for Flow Cytometry
Spezifikation
| B7-H7 | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
| Q9UM44 | |
| HHLA2 | |
| B7-H7 recombinant protein | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| MA57YW | |
| APC | |
| HHLA2 | |
| B7-H5; B7H7; B7-H7; B7y; HERV-H LTR-associating protein 2; HHLA2; Human endogenous retrovirus-H long terminal repeat-associating protein 2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11148 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.