missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Basal Bodies of Cillia Monoclonal antibody specifically detects Basal Bodies of Cillia in Human, Hamster samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
Specifications
| Antigen | Basal Bodies of Cillia |
| Anwendungen | Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Klassifikation | Monoclonal |
| Klon | LhS 28 |
| Konjugat | Biotin |
| Zusammensetzung | PBS |
| Wirtsspezies | Mouse |
| Immunogen | BHK a21 Cytoskeletal preparation |
| Reinigungsverfahren | Protein A purified |
| Menge | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?