missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-XL Mouse anti-Human, Clone: 4C12A6, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Spezifikation
| Antigen | Bcl-XL |
|---|---|
| Klon | 4C12A6 |
| Konzentration | 1 mg/mL |
| Anwendungen | Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
16837525
|
Proteintech
66020-1-IG-20UL |
20 μL |
145.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
16827525
|
Proteintech
66020-1-IG-150UL |
150 μL |
410.00 €
150 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
BCL-XL is a part of the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. BCL-XL is located at the outer mitochondrial membrane, and have been shown to regulate outer mitochondrial membrane channel (VDAC) opening. VDAC regulates mitochondrial membrane potential and controls the production of reactive oxygen species and release of cytochrome C by mitochondria, both of which are the potent inducers of cell apoptosis. Two alternatively spliced transcript variants, which encode distinct isoforms, have been reported. The longer isoform acts as an apoptotic inhibitor and the shorter form acts as an apoptotic activator.Spezifikation
| Bcl-XL | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Apoptosis regulator Bcl X, BCL2L1, Bcl-XL | |
| BCL2L1 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 4C12A6 | |
| Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q07817 | |
| 598 | |
| Bcl-XL Fusion Protein Ag18037 | |
| Primary | |
| -20°C | |
| BCL2L1 |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts