Learn More
BCL11B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 55414-1-AP-20UL
Descripción
This antibody is specific to BCL11B. The calculated molecular weight of BCL11B is 95 kDa, but modified BCL11B protein is about 116 kDa.
This gene encodes a C2H2-type zinc finger protein and is closely related to BCL11A, a gene whose translocation may be associated with B-cell maligcies. The specific function of this gene has not yet been determined. Two alternatively spliced transcript variants, which encode distinct isoforms, have been reported.Especificaciones
| BCL11B | |
| Polyclonal | |
| Unconjugated | |
| BCL11B | |
| B cell CLL/lymphoma 11B, B cell lymphoma/leukemia 11B, BCL11B, COUP TF interacting protein 2, CTIP 2, CTIP2, hRit1, hRIT1 alpha, RIT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 58208, 64919 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99PV8, Q9C0K0 | |
| BCL11B | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.