Learn More
BD Spritze mit Luer-Lock-Spitzen (ohne Nadel)
Artikelnummer. 15578893
Click to view available options
Volumen (metrisch):
10 ml
50 ml
Graduierungen:
1 ml
1-5 ml
NOTICE: The printing has changed on BD 60 mL syringes to only be graduated to 50 mL now. The syringe itself has not changed. Syringes graduated to 60 mL are no longer available.
Artikelnummer. 15578893
Marke: BD 301029
om dit product te kopen Registreer vandaag om een webaccount aan te maken
NOTICE: The printing has changed on BD 60 mL syringes to only be graduated to 50 mL now. The syringe itself has not changed. Syringes graduated to 60 mL are no longer available.
Siliconized and assembled with scale and tip shield
  • Single-use with plastic barrels, plungers and tips
  • Nonsterile syringes may by irradiated, autoclaved, or gas sterilized out of polybag
TRUSTED_SUSTAINABILITY
Spezifikation
Material Kunststoff
Produkttyp Spritze
Graduierungen 1-5 ml
Nummer der Kanülenspitze Ohne Kanüle
Volumen (metrisch) 10 ml
Anschlusstyp Luer-Lock