Learn More
BDNF Polyclonal Antibody, Invitrogen™
Guinea Pig Polyclonal Antibody
Marke: Invitrogen PA5111802
Beschreibung
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)VLEKVPV SKQLK Homology: Mouse, rat and many other species -identical.
BDNF (Brain-Derived Neurotrophic Factor) is a member of the neurotrophin family. BDNF is synthesized as pre-proBDNF, followed by cleavage to proBDNF. Although further processing generates the mature, 14 kDa protein, pro-BDNF is biologically active and is secreted in synaptic vesicles along with the mature form. BDNF is widely expressed in the central nervous system and acts in an autocrine and paracrine manner on several classes of neurons. Signaling occurs mainly through the tyrosine kinase receptor TrkB, although binding to the lower-affinity receptor, p75-NTR, has also been demonstrated. BDNF promotes neuronal survival and differentiation and has been shown to play a critical role in memory formation and synaptic regulation. BDNF is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of BDNF is reduced in both Alzheimer′s and Huntington disease patients, and may also play a role in the regulation of stress response and in the biology of mood disorders.
Spezifikation
| BDNF | |
| Polyclonal | |
| Unconjugated | |
| BDNF | |
| Abrineurin; ANON2; BDNF; BDNF precursor form; Brain-derived neurotrophic factor; BULN2; H-BDNF; ProBDNF | |
| Guinea Pig | |
| Antigen affinity chromatography | |
| RUO | |
| 12064, 24225, 627 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| P21237, P23363, P23560 | |
| BDNF | |
| Peptide corresponding to amino acid residues 166-178 of human BDNF (precursor) | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.