missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Beclin 1 Recombinant Rabbit Monoclonal Antibody (24GB4550)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557348
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Beclin 1 Recombinant Monoclonal Antibody for Western Blot
Beclin 1 plays a central role in autophagy. It acts as a core subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate. Different PI3K complex forms are believed to play a role in multiple membrane trafficking pathways - PI3KC3-C1 is involved in the initation of autophagosomes and PI3KC3-C2 is involved in the maturation of autophagosomes and endocytosis. Beclin 1 also plays a role in antiviral host defense.
Spezifikation
| Beclin 1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| BECN1 | |
| 4921513J16Rik; 5430417M23Rik; APG6; Atg6; ATG6 autophagy related 6 homolog; Bcl-2-interacting protein beclin; beclin 1; beclin 1 (coiled-coil, moesin-like BCL2-interacting protein); beclin 1 (coiled-coil, myosin-like BCL2 interacting protein); beclin 1 (coiled-coil, myosin-like BCL2-interacting protein); beclin 1, autophagy related; beclin1; beclin-1; Beclin-1; beclin-1; Beclin-1-C 35 kDa; Beclin-1-C 37 kDa; becn1; Coiled-coil myosin-like BCL2-interacting protein; coiled-coil, myosin-like BCL2-interacting protein; GT197; hypothetical protein MGC73097; myosin-like BCL2 interacting protein; Protein GT197; testis secretory sperm-binding protein Li 215e; VPS30; zgc:73097 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114558, 56208, 8678 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 24GB4550 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88597, Q14457, Q91XJ1 | |
| BECN1 | |
| Recombinant protein of human Beclin 1 (aa 1-200). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur