Learn More
Benzoylcyanid, TRC
Specifications
Specifications
| Chemischer Name oder Material | Benzoyl Cyanide |
| CAS | 613-90-1 |
| Summenformel | C8H5NO |
| Menge | 5 g |
| InChI-Formel | InChI=1S/C8H5NO/c9-6-8(10)7-4-2-1-3-5-7/h1-5H |
| SMILES | O=C(C#N)c1ccccc1 |
| IUPAC-Name | Benzoylcyanid |
| Molekulargewicht (g/mol) | 131.13 |
| Formelmasse | 131.0371 |