missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ beta-1 Adrenergic Receptor Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 10325094
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
Artikelnummer. Menge unitSize
10325094 100 μL 100 Mikroliter
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 10325094

Marke: Invitrogen™ PA1049

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

PA1-049 detects beta-1 adrenergic receptor (B1AR) from , human, mouse, non-human primate, and rat tissues. PA1-049 has been successfully used in Western blot and immunofluorescence procedures. By Western blot, this antibody detects an ~50.5 kDa band representing B1AR in mouse kidney membrane preparation. Immunofluorescence staining of B1AR in mouse kidney distal tubule with PA1-049 yields a pattern consistent with plasma membrane staining. The PA1-049 immunogen is a synthetic peptide corresponding to residues H(394) G D R P R A S G C L A R A G(408) of mouse B1AR. This sequence is completely conserved between mouse and rat B1AR.

Adrenergic receptors (ARs) are members of the 7-transmembrane domain G-protein-coupled receptor superfamily that bind the endogenous catecholamines epinephrine and norepinephrine. Pharmacological, structural, and molecular cloning data indicate significant heterogeneity within this receptor family. Nine receptor subtypes have been identified thus far including three alpha-1 AR subtypes (1A/D, 1B, and 1C), three alpha-2 ARs (2A, 2B, and 2C), and three beta AR subtypes (1, 2, and 3). ARs participate in either the onset or maintenance of several disease states including hypertension, cardiac dysfunction (congestive heart failure, ischemia, arrhythmias), diabetes, glaucoma, depression, and impotence. BARs participate in diverse processes including development, behavior, cardiac function, smooth muscle tone, and metabolism. In gene-knockout experiments, the majority of mice that lack the B1AR gene die prenatally and those that do survive until adulthood display abnormal cardiac function. Other studies have shown that the direct regulation of cardiac B1AR density by thyroid hormones occurs at the transcriptional level and is modulated by the catecholamine sensitive-adenylyl cyclase system. Evidence for the effects of cAMP on B1AR regulation has come from a study of members of the cAMP response element (CRE) modulator (CREM) family of transcription factors in transformed cell lines.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen beta-1 Adrenergic Receptor
Anwendungen Immunohistochemistry, Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Konzentration Conc. Not Determined
Konjugat Unconjugated
Zusammensetzung Whole serum with 0.05% sodium azide
Gen ADRB1
Gen-Zugriffsnummer P08588, P18090, P34971
Gen-Alias Adrb1; Adrb-1; ADRB1R; adrenergic receptor, beta 1; adrenergic, beta-1-, receptor; adrenoceptor beta 1; B1AR; beta 1-adrenergic receptor beta 1-AR; beta 1-AR; beta-1 adrenergic receptor; Beta-1 adrenoceptor; beta-1 adrenoreceptor; BETA1AR; beta-AR; cardiac beta adrenergic receptor; RATB1AR; RHR
Gensymbole ADRB1
Wirtsspezies Rabbit
Immunogen Synthetic peptide corresponding to residues H(394) G D R P R A S G C L A R A G(408) of mouse B1AR.
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 11554, 153, 24925
Zielspezies Human, Mouse, Rat, Monkey
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.