Beschreibung
The Beta Actin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Beta Actin. This antibody recognizes human, mouse, rat, zebrafish, plant antigen. The Beta Actin antibody has been validated for the following applications: WB analysis. This antibody is hrp.
Spezifikation
Spezifikation
| Antigen | Beta Actin |
| Anwendungen | Western Blot, Immunofluorescence, ELISA |
| Klassifikation | Monoclonal |
| Konzentration | 1000 μg/mL |
| Konjugat | HRP |
| Wirtsspezies | Mouse |
| Menge | 100 μL |
| Primär oder sekundär | Primary |
| Gen-ID (Entrez) | 60 |
| Zielspezies | Human, Mouse, Rat, Zebrafish, Plant |
| Mehr anzeigen |