Learn More
Beta Dystroglycan Antibody (P38G1) - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Marke: Novus Biologicals NBP3-43570
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Beta Dystroglycan Monoclonal antibody specifically detects Beta Dystroglycan in Human,Mouse samples. It is validated for Flow Cytometry,ELISA
Spezifikation
| Beta Dystroglycan | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, ELISA | |
| A3a, AGRNR, DAG156DAG, dystroglycan, dystroglycan 1 (dystrophin-associated glycoprotein 1), Dystrophin-associated glycoprotein 1 | |
| The original antibody was generated by immunizing BALB/c mice with a mixture of transfected HEK 293 cells expressing human and mouse beta-DAG. (Uniprot# Q14118; Q62165) | |
| 0.2 mg | |
| Breast Cancer, Cancer | |
| 1605 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, ELISA | |
| P38G1 | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |