missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bicarbaminsäurediisopropylester, TRC
Specifications
Specifications
| Chemischer Name oder Material | Diisopropyl Bicarbamate |
| CAS | 19740-72-8 |
| Summenformel | C8 H16 N2 O4 |
| Menge | 2.5 g |
| Synonym | Diisopropyl bicarbamate |
| InChI-Formel | InChI=1S/C8H16N2O4/c1-5(2)13-7(11)9-10-8(12)14-6(3)4/h5-6H,1-4H3,(H,9,11)(H,10,12) |
| SMILES | CC(C)OC(=O)NNC(=O)OC(C)C |
| IUPAC-Name | Propan-2-yl-N-(Propan-2-yloxycarbonylamino)carbamat |
| Molekulargewicht (g/mol) | 204.22 |
| Formelmasse | 204.111 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?