Learn More
BID p15 Polyclonal Antibody
BID p15 Polyclonal Antibody
Marke: Invitrogen 44436G
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
BID p15 Polyclonal Antibody for Western Blot, ICC/IF
Spezifikation
| BID p15 | |
| Polyclonal | |
| Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
| P55957, P70444 | |
| BID | |
| The antiserum was produced against a chemically synthesized peptide derived from the 15 kDa caspase-8 cleavage product of mouse BID. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| BID | |
| 2700049M22Rik; AI875481; apoptic d; apoptic death agonist; AU022477; BH3 interacting domain death agonist; BH3-interacting domain death agonist; BH3-interacting domain death agonist p11; BH3-interacting domain death agonist p13; BH3-interacting domain death agonist p15; BID; BID isoform ES(1b); BID isoform L(2); BID isoform Si6; desmocollin type 4; FP497; HGNC:1050; Human BID coding sequence; MGC15319; MGC42355; OTTHUMP00000196197; p11 BID; p13 BID; p15 BID; p22 BID | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12122, 637 | |
| -20°C | |
| Liquid |