Learn More
Ansell BIOCLEAN™ Permaflow Irradiated Cleanroom Ballpoint Pen S-BPBP-1
Click to view available options
Farbe:
Blau
Schwarz
Packungsgröße:
300 Stück
Beschreibung
- BioClean Permaflow cleanroom pens have fade and water resistant fast drying permanent ink, and feature an alcohol resistant label for easy cleaning within a critical environment, they are ISO Class 4 and EU GMP Grade A compatible.
- With a robust, lightweight, tinted polypropylene round barrel and 1 mm point size for quality writing precision. Ink complies with documentary use standard ISO 12757-2
Spezifikation
Spezifikation
| Spitzenform | Rollerball |
| Produkttyp | Ballpoint Pen |
| Farbe | Schwarz |