Learn More
Birch pollen Bet V1 Antibody (M0418) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
Marke: Novus Biologicals NBP3-09012-0.05mg
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Birch pollen Bet V1 Monoclonal antibody specifically detects Birch pollen Bet V1 in Plant samples. It is validated for ELISA, X-Ray Crystallography
Spezifikation
| Birch pollen Bet V1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Human | |
| Affinity purified | |
| RUO | |
| Plant | |
| Purified |
| ELISA, X-Ray Crystallography | |
| M0418 | |
| Unconjugated | |
| Allergen Bet v I-A, Allergen=Bet v 1-A, BETVI, BETVIA, Major pollen allergen Bet v 1-A | |
| Bet v 1-A, a major allergen of the European white birch. | |
| 0.05 mg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgE λ |