missing translation for 'onlineSavingsMsg'
Learn More

BIX, TRC

Product Code. 30332954
Change view
Click to view available options
Menge:
5 mg
Unit Size:
5 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
30332954 5 mg 5 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332954 Supplier Toronto Research Chemicals Supplier No. TRCB5917685MG
 Eingeschränkter Artikel
Verfügbar ab: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material BIX
CAS 101714-41-4
Summenformel C9 H7 N O3 S
Menge 5 mg
Synonym BIX, 2-(3,4-Dihydroxyphenyl)-2-oxoethyl Ester Thiocyanic Acid
InChI-Formel InChI=1S/C9H7NO3S/c10-5-14-4-9(13)6-1-2-7(11)8(12)3-6/h1-3,11-12H,4H2
SMILES Oc1ccc(cc1O)C(=O)CSC#N
IUPAC-Name [2-(3,4-DiHydroxyphenyl)-2-oxoethyl]thiocyanat
Molekulargewicht (g/mol) 209.222
Formelmasse 209.015
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.