missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLNK Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10855-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
B cell linker protein (Blnk) is a protein encoded by the Blnk gene located on chromosome 10 in humans. It is also known as SLP 65 (Src homology 2 domain-containing leukocyte protein of 65 kDa). It derives its name from being an adaptor protein in the B cell receptor signaling cascade with several supporting studies showing improper development of B cells in the absence of Blnk. Blnk undergoes tyrosine phosphorylation by Syk, a B cell receptor activated kinase, which provides a platform for interaction of signaling intermediaries such as the Vav GEF and Grb2.Spezifikation
| BLNK | |
| Polyclonal | |
| Unconjugated | |
| BLNK | |
| B cell linker, B cell linker protein, BASH, BLNK, BLNK S, Cytoplasmic adapter protein, LY57, SLP 65, SLP65 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17060, 29760 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8WV28, Q9QUN3 | |
| Blnk | |
| BLNK Fusion Protein Ag1290 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur