missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMI-1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NB110-40823H
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
BMI-1 Polyclonal specifically detects BMI-1 in Human, Mouse samples. It is validated for Western Blot.
Spezifikation
| BMI-1 | |
| Polyclonal | |
| Western Blot | |
| P35226 | |
| BMI1 | |
| A synthetic peptide made to an internal region (within residues 1-100) of human Bmi1. [Swiss-Prot# P35226] | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Hematopoietic Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators | |
| 648 | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| HRP | |
| PBS with No Preservative | |
| B lymphoma Mo-MLV insertion region 1 homolog, B lymphoma Mo-MLV insertion region 1 homolog (mouse), BMI1 polycomb ring finger oncogene, FLVI2/BMI1, MGC12685, murine leukemia viral (bmi-1) oncogene homolog, PCGF4, polycomb complex protein BMI-1, polycomb group protein Bmi1, polycomb group ring finger 4, RING finger protein 51, RNF51Polycomb group RING finger protein 4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. Immunogen sequence has 92% homology to primate, feline, chicken, bovine, rabbit, rat and Zebrafish protein. | |
| Store at 4C in the dark. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur