Learn More
Thermo Scientific™ Bone Morphogenetic Protein 7, Human, Inactive Protein
Click to view available options
Menge:
10 μg
2 μg
Packungsgröße:
10 Mikrogramm
2 Mikrogramm
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spezifikation
Spezifikation
| Zusammensetzung | Lyophilized powder |
| Menge | 2 μg |
| Quelle | Escherichia coli |
| Lagerungsbedingungen | -30° to -10°C |
| Kennzeichnung | RUO |
| Produkttyp | Bone Morphogenetic Protein 7, Human, inactive protein |
| Rekombinant | Native |
| Form | Lyophilized |