Learn More
uvex Boots S3
Alle ansehen Uvex Produkte
Click to view available options
Größe:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Packungsgröße:
1 Paar
Beschreibung
The uvex 3S3 68742 safety shoe has been specially developed for the extreme demands faced in heavy applications with SR slip resistance.
- It combines necessary robustness with a sporty and dynamic design.
- Equipped with the latest uvex technologies, the uvex 3 reliably protects the wearer while providing high levels of comfort.
- EN ISO 20345: S3L
Spezifikation
Spezifikation
| Farbe | Black, Yellow |
| Material | Leather, Textile |
| Größe | 44 |
| Zehenmaterial | Polymer |
| Einwegprodukt | Reusable |
| Geschlecht | Men's |
| Fit | Standard Width |
| Latexfrei | Yes |
| Waterproof | Yes |
| Produkttyp | Boots |