Learn More
Borsäure-Lösung
Artikelnummer. 11969172
Ansicht ändern
Click to view available options
Menge:
500 ml
Packungsgröße:
500 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
11969172 500 ml 500 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 11969172 Lieferant Chem-Lab Lieferanten-Nr. CL0202070500
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
Spezifikation
Farbe Farblos
Siedepunkt 100 °C
Schmelzpunkt 0 °C
Material 40 g H3BO3/ l H2O
Dichte 1 g/ml
Löslichkeitsinformationen Wasserlöslich
Geruch Geruchsneutral
pH ±4
Menge 500 ml
Physikalische Form Flüssig