missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bovine IL-4 Recombinant Protein

Artikelnummer. 11858761
Click to view available options
Menge:
20 μg
Packungsgröße:
20 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Artikelnummer. 11858761

Marke: Invitrogen™ RBOIL4I

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Recombinant Protein

For dilution, use a buffer containing carrier protein, e.g. PBS with 1% BSA or a neutral media containing 10% FCS and freeze in aliquots at -70°C. Activity: The biological activity of recombinant bovine IL-4 was determined by measuring the proliferation of the factor-dependent human T-helper cell line, TF-1 (Kitamura, T. et al. (1989) J. Cell Physiol. 140:323). The ED50 (or 1 unit of activity) for this effect in our laboratories typically falls into the range of 0.44 - 0.67 ng/mL. Expression: Mature bovine IL-4 was expressed in Pichia pastoris, X-33. GenBank™ Accession # P30367, amino acids 25 - 135. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: HKCDITLAEI. Reconstitute with 0.5 mL of distilled water.

Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P30367
Zur Verwendung mit (Anwendung) ELISA, Immunoprecipitation, Western Blot
Zusammensetzung PBS with no preservative
Gen-ID (Entrez) 280824
Molekulargewicht 12.6 kDa
Name Bovine IL-4
Menge 20 μg
Quelle Yeast
Kennzeichnung RUO
Gen-Alias B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4 delta2; interleukin 4 variant IL-4 delta3; interleukin 4 variant IL-4 int2A; interleukin 4 variant IL-4 int3A; interleukin 4 variant IL-4 int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra
Gebräuchliche Bezeichnung IL-4
Gensymbol IL4
Proteinlänge aa 25-135
Biologische Aktivität ED50 = 0.44 - 0.67 ng/mL; determined by the dose-dependent proliferation of human TF-1 cells.
Konjugat Unconjugated
Spezies Yeast
Rekombinant Recombinant
Inhalt und Lagerung 4°C
Expressionssystem P. pastoris
Form Lyophilized
Protein-Subtyp Recombinant
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt